Tubing with small flanges
Leverandør: VACUUBRAND
20686010 20686011 20686012 20686020 20686021 20686022
189-3310EA 1600 NOK
189-3310 189-3312 189-3314 189-3311 189-3313 189-3315
Tubing with small flanges
Slanger Vakuumslanger
PVC.
  • Resistant to most chemicals
  • Assembly and disassembly are simple and easy
  • Safe, vacuum-tight connection when used with optional centring and clamping rings
Tubing with internal stainless steel spiral and aluminium small flanges mounted on both ends.