Renromsservietter, Quiltec® I
Leverandør: CONTEC
QT1-99 QT1-1212 LWQT0019 QT1-99B QT1-1212B/100 LWQT0034 QT1-66B
115-0378EA 1750 NOK
115-0378 115-3077 115-3075 818-0301 115-3078 115-3076 115-3079
Renromsservietter, Quiltec® I
Papirservietter Renromsservietter Tørkepapir, renrom
Quiltec I wipes offer the highest sorptive capacity of any sealed edge wipe available. They are manufactured using two layers of knitted polyester fabric that are laminated together to form a stronger, more adsorbent wipe.
  • Most sorbent sealed edge wipe available
  • Extremely low in both particles and fibres reducing the possibility of contamination
  • Strong fabric is abrasion and chemical resistant
  • Laser-cut, which seals fibres at wipe's edge
  • Excellent sorbency with solvents
  • Laundered and packed in a Class 4 cleanroom
Additional fluid is captured withinMore Product Information
Bestill nå
SPESIFIKASJONER
Modell Quiltec® I
Environmentally Preferable
grunnvekt 240 g/m²
Absorberingsevne - utvendig 592 ml/m²
Absorberingsevne - innvendig 2,53 ml/g
Absorberingsevne - hastighet <1 s
Ekstraherbare stoffer - bruk av deionisert vann 0,003 g/m²
Ekstraherbare stoffer - bruk av isopropylalkohol 0,010 g/m²
Fibre (>100 µm) 0,11×10³/m²
Partikkelgenerering(>=0.5 µm) 2,66 × 10⁶/m²
Ioner - klorid 0,03 ppm
Ioner - natrium 0,05 ppm