Wipes for general applications, Tork
Leverandør: Essity
510104 510137 510237
115-0640EA 4625 NOK
115-0640 115-0338 115-1658
Wipes for general applications, Tork
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
Strong and durable cloth wipes for use in a variety of environments, softness and flexibility mean wipes are excellent for cleaning, wiping and polishing even in confined spaces.
  • Cellulose/PP/polyester, reusable
  • Solvent resistant, good dry and wet strength and performance
  • High absorbency
  • Low-linting
Farge: hvit eller blå.
Colour: White or blue