Magnifier, Linen Tester
Leverandør: ESCHENBACH OPTIK GMBH
630-8321EA 460 NOK
ESCH12003
Magnifier, Linen Tester
Linen tester with brass frame, satin matt chrome-plated.
  • Biconvex aplanatic lens
  • Additional measuring scale, graduation 1/10 mm
  • Cut-out height: 18 mm Ø 33 mm