Krom ≥99.95% (metals basis), pulver -200 mesh
Leverandør: Thermo Scientific
Warning
13796.14 13796.22 13796.36
13796.14EA 1800 NOK
13796.14 13796.22 13796.36
Krom ≥99.95% (metals basis), pulver -200 mesh
Krom
Purity: 99.95% CAS No.: 7440-47-3 Appearance: Silver Odour: Odourless UN No.: 3089 MDL No.: MFCD00010944
Formel: Cr
MW: 52 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010944
CAS-nummer: 7440-47-3
EINECS: 231-157-5
FN: 3089
ADR: 4.1,III