Mikrosentrifugerør med skrukork, 1,5/2,0 ml, Corning®
Leverandør: Corning
430909 430915 430917
734-1873EA 3450 NOK
734-1873 734-1874 734-1875
Mikrosentrifugerør med skrukork, 1,5/2,0 ml, Corning®
Rør Mikrosentrifugerør
Polypropylene, with orange screw cap.
  • Good chemical resistance
  • Good temperature resistance
  • Screw caps with O-rings provide a tight secure seal
  • With large white marking spot