Saltsyre 2 mol/l (2 N), AVS TITRINORM volumetric solution
Leverandør: VWR Chemicals
Warning
Synonymer: Hydrochloric acid solution, Muriatic acid
30025.293EA 225 NOK
30025.293 30025.362 30025.373 30025.407 30025.320
Saltsyre 2 mol/l (2 N), AVS TITRINORM volumetric solution
Saltsyre
Formel: HCl
MW: 36.46 g/mol
Tetthet: 1.031 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8 III
Limited Time Sale
Select items below are currently on sale
Bestill nå
Spesifikasjon Testresultater
Titer (20°C; real value 0.2 % accuracy) 1.996 - 2.004 mol/l
NIST traceable Confirmed