Laboratory trays
Leverandør: Brand
156640 156642 156644 156646 156648 156650
BRND156640EA 255 NOK
BRND156640 BRND156642 BRND156644 BRND156646 BRND156648 BRND156650
Laboratory trays
Skåler Laboratoriebrett/-skåler
PP, white.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
With ribs on base. Highly versatile trays, ideal for a wide range of applications. The ribs on the base make these trays ideal for use as developing trays.