1-(2-Pyridylazo)-2-naphthol (PAN) ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: PAN, 1-(2-Pyridylazo)-2-naphthol
A17475.03 A17475.06 A17475.14
A17475.03EA 565 NOK
A17475.03 A17475.06 A17475.14
1-(2-Pyridylazo)-2-naphthol (PAN) ≥98%
1-(2-Pyridylazo)-2-naphthol
Purity: 98% CAS No.: 85-85-8 Molecular Formula: C15H11N3O Formula Weight: 249.27 Melting Point: 138-141° Beilstein Reference: 206390
Formel: C₁₅H₁₁N₃O
MW: 249,27 g/mol
Kokepunkt: 412 °C (1013 hPa)
Smeltepunkt: 138…141 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004071
CAS-nummer: 85-85-8
EINECS: 201-637-9