2,4-Dihydroxy-3-nitro-6-picoline ≥97%
Leverandør: Thermo Scientific
Warning
Synonymer: 2,4-Dihydroxy-6-methyl-3-nitropyridine, 4-Hydroxy-6-methyl-3-nitropyridin-2(1H)-one, 4-Hydroxy-6-methyl-3-nitro-2-pyridone
A19690.14 A19690.22
A19690.14EA 845 NOK
A19690.14 A19690.22
2,4-Dihydroxy-3-nitro-6-picoline ≥97%
2,4-Dihydroxy-3-nitro-6-picoline
Purity: 97% CAS No.: 4966-90-9 Molecular Formula: C6H6N2O4 Formula Weight: 170.13 Melting Point: 250° dec. Beilstein Reference: 1529425
Formel: C₆H₆N₂O₄
MW: 170,12 g/mol
Smeltepunkt: >250 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167612
CAS-nummer: 4966-90-9