4-Bromoimidazole ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 4-Bromo-1H-imidazole

B3280-1G B3280-5G
TCIAB3280-1GEA 220 NOK
TCIAB3280-1G TCIAB3280-5G
4-Bromoimidazole ≥98.0% (by GC, titration analysis)
4-Bromoimidazole
Formel: C₃H₃BrN₂
MW: 146,97 g/mol
MDL nummer: MFCD00047021
CAS-nummer: 2302-25-2
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4efc13fc6dd4b7eb0655dce88adb932afe9506d8&vid=4efc13fc6dd4b7eb0655dce88adb932afe9506d8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR