2-Naftyl benzoat ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Benzoic acid 2-naphthyl ester

B1108-25G B1108-500G
TCIAB1108-25GEA 365 NOK
TCIAB1108-25G TCIAB1108-500G
2-Naftyl benzoat ≥98.0% (by GC)
2-Naftyl benzoat
Formel: C₁₇H₁₂O₂
MW: 248,28 g/mol
Kokepunkt: 414 °C (1013 hPa)
Smeltepunkt: 104…106 °C
Tetthet: 1,1136 g/cm³ (110 °C)
MDL nummer: MFCD00014320
CAS-nummer: 93-44-7
EINECS: 202-247-1
Merck Indeks: 14,06408
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc53e4c0b87fc00f3b34704fed60ba890509bb36&vid=04675feb6fd58073965fdbdd889cc0d3a13e5ac3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR