Butanon, EMPLURA®, Supelco®
Leverandør: Merck
Danger
Synonymer: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
M-Clarity™ quality level = MQ200
1.06014.2500 1.06014.6025 1.06014.9190 1.06014.9011
1.06014.2500EA 1325 NOK
1.06014.2500 1.06014.6025 1.06014.9190 1.06014.9011
Butanon, EMPLURA®, Supelco®
Methyl ethyl ketone
Formel: C₂H₅COCH₃
MW: 72,11 g/mol
Kokepunkt: 79,6 °C (1013 hPa)
Smeltepunkt: –86 °C
Tetthet: 0,8054 g/cm³ (20 °C)
Flash punkt: –4 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011648
CAS-nummer: 78-93-3
EINECS: 201-159-0
FN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Indeks: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM