Urglass
Leverandør: Brand
150904 150907 150910 150912 150915 150920
BRND150904EA 210 NOK
BRND150904 BRND150907 216-1155 BRND150912 BRND150915 BRND150920
Urglass
Urglass
PTFE, white.
  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth surfaces to prevent contamination
Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.