Butylacetat ≥99% semiconductor grade
Leverandør: Thermo Scientific
|
Warning
|
19395.K2
19395.K7
19395.M1
19395.K2EA
870
NOK
19395.K2
19395.K7
19395.M1
Butylacetat ≥99% semiconductor grade
Butylacetat
Purity: 99% min
CAS No.: 123-86-4
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 25°(77°F)
Physical Form: Liquid
Density: 0.881
Refractive Index: 1.3945
Appearance: Colourless
Odour: Fruit-like
pH: 5
Melting Point: -78°
Boiling Point: 124-126°
Beilstein Reference: 1741921
Merck Reference: 14,1535
UN No.: 1123
MDL No.: MFCD00009445
Formel:
CH₃COO(CH₂)₃CH₃ Kokepunkt: 124…126 °C Smeltepunkt: –78 °C Tetthet: 0,881 Flash punkt: 25 °C (77 °F) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00009445 CAS-nummer: 123-86-4 EINECS: 204-658-1 FN: 1123 ADR: 3,III Merck Indeks: 14,1535 |