Filterpapir for bruksområder i helsevesenet, Whatman™
Leverandør: Whatman products (Cytiva)
10318489 10318493
WHAT10318489EA 1325 NOK
WHAT10318489 WHAT10318493
Filterpapir for bruksområder i helsevesenet, Whatman™
Filter Filterpapir Kvalitativt filterpapir
These filter papers can be used for the absorption of culture media, as a blotting paper, for electrophoresis and amino acid chromatography.
  • Soft surface
  • For gelatinous samples