2-Propanethiol for syntese, Sigma-Aldrich®
Leverandør: Merck
|
Danger
|
Synonymer:
Isopropyl mercaptan, Isopropanethiol, 2-Mercaptopropane
M-Clarity™ quality level = MQ200
8.07532.0100
8.07532.0500
8.07532.9045
8.07532.0100EA
760
NOK
8.07532.0100
8.07532.0500
8.07532.9045
2-Propanethiol for syntese, Sigma-Aldrich®
2-Propanethiol
Formel:
C₃H₈S MW: 76,16 g/mol Kokepunkt: 57 °C (1013 hPa) Smeltepunkt: –130 °C Tetthet: 0,81 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004863 CAS-nummer: 75-33-2 EINECS: 200-861-4 FN: 2402 ADR: 3,II |
Spesifikasjon Testresultater
Solubility | (20 °C) slightly soluble |
Melting Point | -130 °C |
Molar Mass | 76.16 g/mol |
Boiling Point | 51 - 53 °C (1013 hPa) |
Vapor Pressure | 605 hPa (37 °C) |
Flash Point | -34 °C |
Refractive Index | 1.4255 (20 °C, 589 nm) |
Density | 0.81 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...