n-Trikosan ≥99%

Leverandør: Thermo Scientific

Synonymer: Tricosane

A14577.06 A14577.14 A14577.22
A14577.06EA 1025 NOK
A14577.06 A14577.14 A14577.22
n-Trikosan ≥99%
n-Trikosan

Purity: 99% CAS No.: 638-67-5 Molecular Formula: CH3(CH2)21CH3 Formula Weight: 324.64 Flash Point: 110°(230°F) Refractive Index: 1.4468 Melting Point: 47-50° Boiling Point: 199-200°/3mm Beilstein Reference: 1756000 MDL No.: MFCD00009350

Formel: H₃C(CH₂)₂₁CH₃
MW: 324,63 g/mol
Kokepunkt: 199…200 °C (3 mmHg)
Smeltepunkt: 47…50 °C
Tetthet: 0,7969
Flash punkt: >110 °C (230 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009350
CAS-nummer: 638-67-5
EINECS: 211-347-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR