4-Metyl-2-pentanon 99+% for HPLC
Leverandør: Thermo Scientific
|
Danger
|
43170.K2
43170.K7
43170.M1
43170.M6
43170.K2EA
1375
NOK
43170.K2
43170.K7
43170.M1
43170.M6
4-Metyl-2-pentanon 99+% for HPLC
4-Metyl-2-pentanon
Purity: 99+%
CAS No.: 108-10-1
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 13°(55°F)
Physical Form: Liquid
Density: 0.801
Refractive Index: 1.3960
Odour: Mild
Melting Point: -84°
Boiling Point: 117-118°
Beilstein Reference: 605399
Merck Reference: 14,5207
UN No.: 1245
MDL No.: MFCD00008938
Formel:
(CH₃)₂CHCH₂COCH₃ MW: 100,16 g/mol Kokepunkt: 117…118 °C Smeltepunkt: –84 °C Tetthet: 0,801 Flash punkt: 13 °C (55 °F) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00008938 CAS-nummer: 108-10-1 EINECS: 203-550-1 FN: 1245 ADR: 3,II Merck Indeks: 14,5207 |