Maskeringstaper, krepp, Scotch®
Leverandør: 3M
2021850 2022450
819-0065EA 190 NOK
819-0065 MMMA2022450
Maskeringstaper, krepp, Scotch®
Tape
Scotch® crepe masking tape is a high adhesion tape suitable for masking applications found in new and remedial construction.
  • Conformability that will maintain shape when formed around corners
  • Backing saturated for easy tear, resists slivering yet hand tear able
  • Rubber adhesive provides proper adhesion and ideal holding power
  • Short-term temperature resistance up to 120 °C
Bruk kun på overflater som ikke kan ta skade.
Length: 50,0 m
Colour: Light beige