Acrolein diethyl acetal ≥97.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene
A0640-25G
A0640-250G
TCIAA0640-25GEA
525
NOK
TCIAA0640-25G
TCIAA0640-250G
Acrolein diethyl acetal ≥97.0% (by GC)
Acrolein diethyl acetal
Formel:
C₇H₁₄O₂ MW: 130,19 g/mol Kokepunkt: 124°C Smeltepunkt: –77 °C Tetthet: 0,8543 g/cm³ (15 °C) Flash punkt: 15°C |
MDL nummer:
MFCD00009239 CAS-nummer: 3054-95-3 EINECS: 221-276-0 FN: 2374 ADR: 3,II |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...