1,2,4,5-Tetrabrombensen ≥94%
Leverandør: Thermo Scientific
|
Warning
|
L13539.09
L13539.18
L13539.09EA
600
NOK
L13539.09
L13539.18
1,2,4,5-Tetrabrombensen ≥94%
1,2,4,5-Tetrabrombensen
Purity: 94%
CAS No.: 636-28-2
Molecular Formula: C6H2Br4
Formula Weight: 393.70
Melting Point: 175-180°
Beilstein Reference: 1365830
Formel:
C₆H₂Br₄ MW: 393,7 g/mol Smeltepunkt: 175…180 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00000063 CAS-nummer: 636-28-2 EINECS: 211-253-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...