Trykkslanger med høy renhet, Tygon® 2475, I.B.
Leverandør: Saint Gobain Life Sciences
ACX00029 ACX00019
VERNACX00029EA 11900 NOK
VERNACX00029 VERNACX00019
Trykkslanger med høy renhet, Tygon® 2475, I.B.
Slanger Overføringsslange
Tygon® tubing, clear.
  • Extremely low sorption to aqueous fluids
  • Plasticizer free
  • Tough braid reinforcement for elevated working pressures
High-purity pressure tubing is hydrophobic and resists the sorption (absorption/adsorption) of aqueous fluids.Typical applications include deionized water systems, pharmaceutical, cosmetic processing, sterile fill lines and chemical transfer.
Temperature resistance: –78 to +52 °C
Hardness (Shore durometer A): 72
Density: 0,90
Sertifiseringer: Meets USP Class VI and FDA criteria
Bestill nå
SPESIFIKASJONER
Bøyeradius 25,4 mm 9,5 mm