Kolber, rund bunn, bred åpning, med valset kant
Leverandør: Lenz Laborglas GmbH & CO.KG
07035028 07035037 07035049 07035058 07035070 07035073 07035079 07035088
LENZ07035028EA 3150 NOK
LENZ07035028 LENZ07035037 LENZ07035049 LENZ07035058 LENZ07035070 LENZ07035073 LENZ07035079 LENZ07035088
Kolber, rund bunn, bred åpning, med valset kant
Kolber Rundkolbe
DURAN® Borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • USP Standard
Marked with capacity and a white labelling area.
Sertifiseringer: DIN ISO 24450