Oppløste oksygenprober, InLab® OptiOx™
Leverandør: Mettler - Toledo
51344621 51344623 51344622
664-0075EA 18480 NOK
664-0075 664-0077 664-0076
Oppløste oksygenprober, InLab® OptiOx™
Elektroder Oksygenelektrode
The InLab® OptiOx optical dissolved oxygen sensor is based on the reliable RDO® (Rugged Dissolved Oxygen) technology. RDO® is fast, precise and produces extremely reproducible results.
  • Stable results and rapid response time
  • Immediately ready for use
  • Extremely easy handling, zero maintenance: save time
  • Suitable for an extremely wide range of applications
The InLab® OptiOx sensors can be used in combination with a SevenExcellence benchtop meter or the SevenGo 9 or SevenGo Duo 98 portable meters.