Syringaldehyd

Leverandør: Molekula

Synonymer: 3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Hydroxy-3,5-dimethoxybenzaldehyde

49805673-25G 49805673
MOLE49805673-25GEA 0 NOK
MOLE49805673-25G MOLE49805673
Syringaldehyd
Syringaldehyd
Formel: C₉H₁₀O₄
MW: 182,18 g/mol
Kokepunkt: 322 °C (1013 hPa)
Smeltepunkt: 107 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006943
CAS-nummer: 134-96-3
EINECS: 205-167-5
Merck Indeks: 12,09191

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR