Examination gloves, vinyl, Evercare®
Leverandør: Onemed Sverige AB
295821/6351 295820/6350 295822/6352 295823/6353 295824/6354
113-7909EA 215 NOK
113-7909 113-7908 113-7910 113-7911 113-7912
Examination gloves, vinyl, Evercare®
Hansker
Disposable, powder-free examination gloves for non sterile environments. Ideal for general purposes as well as for food handling (Not suitable for fatty foodstuffs).
  • Non sterile, powder-free
  • Phthalate-free: avoids health concerns
  • Ambidextrous, with rolled rim
  • No latex proteins, good alternative for those suffering from Type I allergies
  • Very soft, good fit, feel and performance
  • In easy-to-use dispenser box of 100 gloves
Colour: Transparent
Length: 240 mm
EN 455; AQL 1,5 Food contact approved
Sertifiseringer: EN 455; AQL 1,5
Food contact approved.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Ambidekstral/håndspesifikk Ambidextrous
Engangs/flergangs engangs
Latex/latexfri Latex-fri
Pudder/pudderfri uten pudder
Overflate Smooth
Type mansjett Perlebesatt
Tykkelse finger 0,08 mm
Mansjett tykkelse 0,08 mm
EN norm EN 455
Godkjent for kontakt med matvarer +