Målekolber, klasse B, rød gradering
Leverandør: KARTELL
2490 2491 2492 2493 2494
KART2490EA 535 NOK
KART2490 KART2491 KART2492 KART2493 KART2494
Målekolber, klasse B, rød gradering
Kolber Målekolber
PMP, gjennomsiktige, med skrukork.
  • Autoclavable to 170 °C
  • Chemically cleaner than glass and chemically non absorbant
  • No wetting
  • Suitable for foodstuff
These crystal clear volumetric flasks create no meniscus so final readings are easy. Calibrated gravimetrically at 20 °C. Thick walls eliminate distortion. Thin necks make final measurement precise.