Blood collection needles, BD Vacutainer®
Leverandør: BD Medical
367300 360214 360212 360211 360210 360213 360215 360213UK 360210UK 367300UK 360214UK
613-3971EA 995 NOK
613-3971 613-3959 613-3957 613-3956 613-3955 613-0832 613-0833 BDAM360213UK BDAM360210UK BDAM367300UK BDAM360214UK
Blood collection needles, BD Vacutainer®
Sprøytenåler
BD Vacutainer® blood collection needles have an advanced low-angle bevel design and are coated with silicone, a low friction lubricant, ensuring very gentle and smooth vein entry.
  • Individual laser quality control maintains the highest quality levels
  • Suitable for use with multiple tube samples
BD Vacutainer® Luer adapters are sterile devices with a multi-sampling valve and are designed for use with a catheter or hypodermic needle to collect blood with blood collection tubes. They are also ideal for use with any female Luer fitting.