6-Chloroanthranilic acid 99%
Leverandør: Thermo Scientific
Warning
Synonymer: 2-Amino-6-chlorobenzoic acid
300500010 300500100
ACRO300500010EA 450 NOK
ACRO300500010 ACRO300500100
6-Chloroanthranilic acid 99%
6-Chloroanthranilic acid
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Kokepunkt: 330 °C (1013 hPa)
Smeltepunkt: 143…147 °C
MDL nummer: MFCD00051530
CAS-nummer: 2148-56-3
EINECS: 218-416-8
Bestill nå
Spesifikasjon Testresultater
Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 147 °C
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %