5-klorvanillin syre tech. 90%, Technical Grade
Leverandør: Thermo Scientific
|
Warning
|
Synonymer:
3-Chloro-4-hydroxy-5-methoxybenzoic acid
A12841.06
A12841.14
A12841.06EA
840
NOK
A12841.06
A12841.14
5-klorvanillin syre tech. 90%, Technical Grade
5-klorvanillin syre
Purity: 90%
CAS No.: 62936-23-6
Molecular Formula: C8H7ClO4
Formula Weight: 202.60
Melting Point: 242-248°
Beilstein Reference: 2648897
Formel:
C₈H₇ClO₄ MW: 202,59 g/mol Smeltepunkt: 242…248 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00016531 CAS-nummer: 62936-23-6 EINECS: 263-766-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...