2,5-Dichlorophenol ≥98%
Leverandør: Thermo Scientific
Warning
B24787.14 B24787.22 B24787.36
B24787.14EA 385 NOK
B24787.14 B24787.22 B24787.36
2,5-Dichlorophenol ≥98%
2,5-Dichlorophenol
Purity: 98% CAS No.: 583-78-8 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 55-58° Boiling Point: 211° Beilstein Reference: 1907692 UN No.: 2020 MDL No.: MFCD00002174
Formel: C₆H₄Cl₂O
MW: 163 g/mol
Kokepunkt: 211 °C
Smeltepunkt: 55…58 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002174
CAS-nummer: 583-78-8
EINECS: 209-520-4
FN: 2020
ADR: 6.1,III