4-Fluorbensonitril ≥99%

Leverandør: Thermo Scientific
Warning

Synonymer: p-Fluorobenzonitrile

A13028.06 A13028.14 A13028.22
A13028.06EA 545 NOK
A13028.06 A13028.14 A13028.22
4-Fluorbensonitril ≥99%
4-Fluorbensonitril

Purity: 99% CAS No.: 1194-02-1 Molecular Formula: C7H4FN Formula Weight: 121.11 Flash Point: 65°(149°F) Odour: Almond-like Boiling Point: 186-188° Beilstein Reference: 2041517 MDL No.: MFCD00001812

Formel: C₇H₄FN
MW: 121,11 g/mol
Kokepunkt: 186…188 °C
Smeltepunkt: 34…37 °C
Tetthet: ∼1 g/cm³ (20 °C)
Flash punkt: 65 °C (149 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001812
CAS-nummer: 1194-02-1
EINECS: 214-784-9
FN: 3439
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR