Screw caps with hose connections, DURAN®
Leverandør: DWK Life Sciences
1129825 293102807
SCOT1129825EA 1100 NOK
SCOT1129825 SCOT293102807
Screw caps with hose connections, DURAN®
Lokk Flaskelokk
PP.
  • Resistant to temperature up to +140 °C
  • Fully autoclavable and washer-proof
  • Suitable for tubing with an inner diamater of 6 to 9 mm
EPDM sealing O-ring for a tight seal with GL 45 bottle.