2-Nitrofenol
Leverandør: Molekula
MOLE28453765-1KGEA
2700
NOK
MOLE28453765-1KG
2-Nitrofenol
2-Nitrofenol
Formel:
C₆H₅NO₃ MW: 139,11 g/mol Kokepunkt: 214,6 °C (1013 hPa) Smeltepunkt: 43…45 °C Tetthet: 1,599 g/cm³ (20 °C) Flash punkt: 108 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00011688 CAS-nummer: 88-75-5 EINECS: 201-857-5 FN: 1663 ADR: 6.1,III Merck Indeks: 12,06717 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...