1-Ethyl-3-methylimidazolium hydrogen sulphate ≥98%
Leverandør: Thermo Scientific
Danger
Synonymer: 1-Ethyl-3-methylimidazolium Hydrogen sulphate
H27232.18 H27232.30
H27232.18EA 1950 NOK
H27232.18 H27232.30
1-Ethyl-3-methylimidazolium hydrogen sulphate ≥98%
1-Ethyl-3-methylimidazolium hydrogen sulphate
Purity: 98% CAS No.: 412009-61-1 Molecular Formula: C6H12N2O4S Formula Weight: 208.24 UN No.: 3265 MDL No.: MFCD06798195
Formel: C₆H₁₂N₂O₄S
MW: 208,24 g/mol
Tetthet: 1,367
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06798195
CAS-nummer: 412009-61-1
FN: 3265
ADR: 8,II