DL-α-Tocopherol acetate, liquid
Leverandør: MP Biomedicals
Synonymer: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
0210055980 0210055905 0210055925
ICNA0210055980EA 2375 NOK
ICNA0210055980 ICNA0210055905 ICNA0210055925
DL-α-Tocopherol acetate, liquid
DL-α-Tocopherol acetate
Tocopherol acetate has properties similar but not identical to α-tocopherol.
Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens.
Freely soluble in absolute ethanol, acetone, chloroform (100 mg/ml - clear solution), ether and fixed oils, soluble in ethanol (96%); slightly soluble in alcohol; practically insoluble in water.
Formel: C₃₁H₅₂O₃
MW: 472.75 g/mol
MDL nummer: MFCD00072042
CAS-nummer: 7695-91-2
EINECS: 231-710-0