4-etylbensaldehyd 98%

Leverandør: Thermo Fisher Scientific
Warning

378950050 378950250 378951000
ACRO378950050EA 275 NOK
ACRO378950050 ACRO378950250 ACRO378951000
4-etylbensaldehyd 98%
4-etylbensaldehyd
Formel: C₉H₁₀O
Kokepunkt: 221 °C (760 mmHg)
Tetthet: 0,97 g/cm³
Flash punkt: 92 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006956
CAS-nummer: 4748-78-1
EINECS: 225-268-8

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5385 to 1.5405 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR