Isobutyrfenon ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Isopropyl phenyl ketone, 2-Methyl-1-phenyl-1-propanone, Phenyl isopropyl ketone, α-Methylpropiophenone
I0113-25ML
I0113-500ML
TCIAI0113-25MLEA
230
NOK
TCIAI0113-25ML
TCIAI0113-500ML
Isobutyrfenon ≥98.0% (by GC)
Isobutyrfenon
Formel:
(C₆H₅)C(O)CH(CH₃)₂ MW: 148,2 g/mol Kokepunkt: 215°C Smeltepunkt: 1 °C Tetthet: 0,98 g/cm³ (20 °C) Flash punkt: 84°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00008917 CAS-nummer: 611-70-1 EINECS: 210-275-0 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...