Silikongummiforseglinger med åpning
Leverandør: DWK Life Sciences
292350607 292361004 292361201 292361604 292361801 292371202 292370806 292340606 292350804 292351003 292361407 292371005 292382602 292383204
SCOT292350607EA 92 NOK
SCOT292350607 275-1338 SCOT292361201 SCOT292361604 SCOT292361801 SCOT292371202 SCOT292370806 SCOT292340606 SCOT292350804 SCOT292351003 SCOT292361407 SCOT292371005 SCOT292382602 SCOT292383204
Silikongummiforseglinger med åpning
Forseglinger Tetningsringer
VMQ with bonded PTFE face.
  • Temperature resistant from −200 to +250 °C
  • Very good chemical resistance
  • Easy handling
For threaded connection caps with hole and GL thread.