Tubing connectors, straight, conical nozzles
Leverandør: BURKLE
8700-0406 8700-0608 8700-0810 8700-1012 8700-1214 8700-1416
BURK8700-0406EA 235 NOK
BURK8700-0406 BURK8700-0608 BURK8700-0810 BURK8700-1012 BURK8700-1214 BURK8700-1416
Tubing connectors, straight, conical nozzles
Fittings og kontakter
Made from PP, white.
  • Sterilisable
  • Autoclavable up to max. 121 °C (20 min.)
  • Suitable for use with foodstuffs
  • Good chemical resistance
Conical, straight hose connectors for various hose sizes.