1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: 1,4-Cyclohexanedicarboxylic acid
B23125.22 B23125.36
B23125.22EA 765 NOK
B23125.22 B23125.36
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) ≥98%
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)
Purity: 98% CAS No.: 1076-97-7 Molecular Formula: C8H12O4 Formula Weight: 172.18 Flash Point: 235°(455°F) Melting Point: 300° Beilstein Reference: 1870377 MDL No.: MFCD00001465
Formel: C₈H₁₂O₄
MW: 172,18 g/mol
Smeltepunkt: >300 °C
Flash punkt: 235 °C (455 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001465
CAS-nummer: 1076-97-7
EINECS: 214-068-6