Jerricans, narrow neck, HDPE, transparent or blue
Leverandør: BURKLE
1427-0005 1427-0010 1427-0020 1427-0030 1427-0002
216-1410EA 205 NOK
216-1410 216-1416 216-1424 216-1426 BURK1427-0002
Jerricans, narrow neck, HDPE, transparent or blue
Flasker Jerrikanne
HDPE, translucent or blue, with tamper evident screw cap.
  • Good chemical resistance
  • Temperature resistant from –40 to +110 °C
  • Can be fitted with screw caps with spigots (optional)
  • Suitable for use with foodstuffs
Universal jerrican for industry, trade and laboratories. Ideal for packing, storage and transfer of liquids.