Screw caps for screw thread tubes
Leverandør: Lenz Laborglas GmbH & CO.KG
01320018 01320014 01320025 01320032 01320045
LENZ01320018EA 82 NOK
LENZ01320018 LENZ01320014 LENZ01320025 LENZ01320032 LENZ01320045
Screw caps for screw thread tubes
Lokk Lokk til reagensglass
PBT,red, with PTFE coated silicone sealing (VMQ).
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air