6-Methylquinoline 98%

Leverandør: Thermo Scientific
Warning

295141000 295140250 295140050
ACRO295141000EA 2750 NOK
ACRO295141000 ACRO295140250 ACRO295140050
6-Methylquinoline 98%
6-Methylquinoline
Formel: C₁₀H₉N
MW: 143,19 g/mol
Kokepunkt: 256…260 °C (760 mmHg)
Smeltepunkt: -22,2 °C
Tetthet: 1,06 g/cm³
Flash punkt: >113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006804
CAS-nummer: 91-62-3
EINECS: 202-084-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to light orange Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR