3-Picolyl chloride hydrochloride 99%
Leverandør: Thermo Scientific
Danger
Synonymer: 3-(Chloromethyl)-pyridinium chloride, 3-(Chloromethyl)pyridine hydrochloride
131190250 131190050
ACRO131190250EA 2400 NOK
ACRO131190250 ACRO131190050
3-Picolyl chloride hydrochloride 99%
3-Picolyl chloride hydrochloride
Formel: C₆H₇Cl₂N
MW: 164,03 g/mol
Smeltepunkt: 137…143 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012818
CAS-nummer: 6959-48-4
EINECS: 230-150-4
FN: 3261
ADR: 8,III
Bestill nå
Spesifikasjon Testresultater
Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥99.4 %
Color scale ≤20 APHA