4-Picolyl chloride hydrochloride ≥98%
Leverandør: Thermo Scientific
Danger
Synonymer: 4-(Chloromethyl)pyridine hydrochloride, 4-(Chloromethyl)-pyridinium chloride
A12859.06 A12859.14 A12859.22
A12859.06EA 490 NOK
A12859.06 A12859.14 A12859.22
4-Picolyl chloride hydrochloride ≥98%
4-Picolyl chloride hydrochloride
Purity: 98% CAS No.: 1822-51-1 Molecular Formula: C6H62N.HCl Formula Weight: 164.04 Melting Point: 166-171° Beilstein Reference: 3689154 UN No.: 3261 MDL No.: MFCD00012826
Formel: C₆H₇Cl₂N
MW: 164,03 g/mol
Smeltepunkt: 166…171 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012826
CAS-nummer: 1822-51-1
EINECS: 217-350-7
FN: 3261
ADR: 8,III