Trifluoromethanesulphonamide 96%

Leverandør: Thermo Fisher Scientific
Warning

B22112.03 B22112.06
B22112.03EA 1350 NOK
B22112.03 B22112.06
Trifluoromethanesulphonamide 96%
Trifluoromethanesulphonamide

Purity: 96% CAS No.: 421-85-2 Molecular Formula: CF3SO2NH2 Formula Weight: 149.09 Melting Point: 116-119° Beilstein Reference: 1812099

Formel: CH₂F₃NO₂S
Smeltepunkt: 116…119 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00068714
CAS-nummer: 421-85-2
EINECS: 431-270-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR