4-Formylphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 4-Boronobenzaldehyde, 4-Formylbenzeneboronic acid, (4-Formylphenyl)metaboric acid, Dihydroxy(4-formylphenyl)borane

F0446-1G F0446-5G
TCIAF0446-1GEA 325 NOK
TCIAF0446-1G TCIAF0446-5G
4-Formylphenylboronic acid (contains varying amounts of Anhydride)
4-Formylphenylboronic acid
Formel: C₇H₇BO₃
MW: 149.94 g/mol
MDL nummer: MFCD00151823
CAS-nummer: 87199-17-5
EINECS: 438-670-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ea94e9f9686e508623cf0d47dec670dc9e32f93&vid=1ea94e9f9686e508623cf0d47dec670dc9e32f93&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 97.0 and le 114.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR