natrium dicyanoamid 96%

Leverandør: Thermo Scientific
Warning

Synonymer: Sodium dicyanamide

B21634.14 B21634.22 B21634.36
B21634.14EA 965 NOK
B21634.14 B21634.22 B21634.36
natrium dicyanoamid 96%
natrium dicyanoamid

Purity: 96% CAS No.: 1934-75-4 Molecular Formula: NaN(CN)2 Formula Weight: 89.03 Melting Point: 300° MDL No.: MFCD00003535

Formel: C₂N₃Na
MW: 89,03 g/mol
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003535
CAS-nummer: 1934-75-4
EINECS: 217-703-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR